accessibility menu, dialog, popup

UserName

CH3H3CCH3BrHOH3CH3CCH3

CAS RN 1139-52-2

IUPAC Name: 4-bromo-2,6-di-tert-butylphenol
Synonyms: 4-bromo-2,6-di-tert-butylphenol
Molecular Weight (g/mol): 285.23
Molecular Formula: C14H21BrO
InChi Key: SSQQUEKFNSJLKX-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(Br)=CC(=C1O)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results