missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,5-dichlorophenol 98.0+%, TCI America™
Supplier: TCI America B2822500G
Specifications
| 4-Bromo-2,5-dichlorophenol | |
| 73°C | |
| C6H3BrCl2O | |
| HWWKEEKUMAZJLL-UHFFFAOYSA-N | |
| 4-bromo-2,5-dichlorophenol | |
| 16022 | |
| ≥98.0% (GC) |
| 1940-42-7 | |
| 187°C | |
| 500 g | |
| C1=C(C(=CC(=C1Cl)Br)Cl)O | |
| 241.893 | |
| 241.89 | |
| Crystalline Powder |
Chemical Identifiers
| 1940-42-7 | |
| 241.893 | |
| 16022 | |
| C1=C(C(=CC(=C1Cl)Br)Cl)O |
| C6H3BrCl2O | |
| HWWKEEKUMAZJLL-UHFFFAOYSA-N | |
| 4-bromo-2,5-dichlorophenol |
Safety and Handling
EINECSNumber : (3)-0956
RTECSNumber : SJ8000000
TSCA : Yes