accessibility menu, dialog, popup

UserName

NaOHClBrCl

CAS RN 1940-42-7

IUPAC Name: 4-bromo-2,5-dichlorophenol sodium
Synonyms: 2,5-dichloro-4-bromophenol sodium
Molecular Weight (g/mol): 264.88
Molecular Formula: C6H3BrCl2NaO
InChi Key: FYRVMZSKWATEMP-UHFFFAOYSA-N
SMILES: [Na].OC1=CC(Cl)=C(Br)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results