missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,5-dichlorophenol 98.0+%, TCI America™

Catalog No. B282225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B282225G 25 g
B2822500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B282225G Supplier TCI America Supplier No. B282225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1940-42-7
Molecular Formula C6H3BrCl2O
Molecular Weight (g/mol) 241.893
InChI Key HWWKEEKUMAZJLL-UHFFFAOYSA-N
PubChem CID 16022
IUPAC Name 4-bromo-2,5-dichlorophenol
SMILES C1=C(C(=CC(=C1Cl)Br)Cl)O

Specifications

Melting Point 73°C
Boiling Point 187°C
Quantity 25 g
Formula Weight 241.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,5-dichlorophenol
EINECSNumber (3)-0956
RTECSNumber SJ8000000
TSCA Yes