missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,3-difluorobenzaldehyde, 98%

Catalog No. AAH6404403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6404403 1 g
AAH6404406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6404403 Supplier Thermo Scientific Chemicals Supplier No. H6404403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 644985-24-0 | C7H3BrF2O | 221.001 g/mol

4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,3-difluorobenzaldehyde is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 644985-24-0
Molecular Formula C7H3BrF2O
Molecular Weight (g/mol) 221.001
MDL Number MFCD09258953
InChI Key IMOLPSNRLZLWQR-UHFFFAOYSA-N
Synonym 2,3-difluoro-4-bromobenzaldehyde, benzaldehyde, 4-bromo-2,3-difluoro, 1-bromo-2,3-difluoro-4-formylbenzene, pubchem10068, 4-bromo-2,3-difluoro-benzaldehyde, 4-bromanyl-2,3-bis fluoranyl benzaldehyde, 4-bromo-2, 3-difluorobenzaldehyde
PubChem CID 26986390
IUPAC Name 4-bromo-2,3-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1C=O)F)F)Br

Specifications

Melting Point 66°C to 67°C
Color White
Quantity 1 g
Beilstein 9551272
Solubility Information Slightly soluble in water.
Formula Weight 221
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Bromo-2,3-difluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only