accessibility menu, dialog, popup

UserName

CAS RN 644985-24-0

IUPAC Name: 4-bromo-2,3-difluorobenzaldehyde
Synonyms: 4-bromo-2,3-difluorobenzaldehyde
Molecular Weight (g/mol): 221
Molecular Formula: C7H3BrF2O
InChi Key: IMOLPSNRLZLWQR-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC(C=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results