missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,3,5,6-tetrafluorobenzotrifluoride, 99%

Catalog No. p-7025710
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2135003 1 g
AAB2135006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2135003 Supplier Thermo Scientific Chemicals Supplier No. B2135003
Only null left

CAS: 17823-46-0 | C7BrF7 | 296.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17823-46-0
Molecular Formula C7BrF7
Molecular Weight (g/mol) 296.97
MDL Number MFCD00075240
InChI Key DONVNRFILRLHJB-UHFFFAOYSA-N
Synonym 1-bromo-2,3,5,6-tetrafluoro-4-trifluoromethyl benzene, 4-bromo-2,3,5,6-tetrafluorobenzotrifluoride, p-bromoheptafluorotoluene, 4-trifluoromethyl-2,3,5,6-tetrafluorobromobenzene, acmc-209ee7, 4-bromoheptafluorotoluene, 4-bromo-alpha,alpha,alpha,2,3,5,6-heptafluorotoluene, 1-trifluoromethyl-2,3,5,6-tetrafluoro-4-bromobenzene
PubChem CID 626637
IUPAC Name 1-bromo-2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene
SMILES C1(=C(C(=C(C(=C1F)F)Br)F)F)C(F)(F)F

Specifications

Density 1.929
Boiling Point 152°C to 153°C
Quantity 1 g
Formula Weight 296.98
Percent Purity 99%
Chemical Name or Material 4-Bromo-2,3,5,6-tetrafluorobenzotrifluoride
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only