accessibility menu, dialog, popup

UserName

FFFFBrFFF

CAS RN 17823-46-0

IUPAC Name: 1-bromo-2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene
Synonyms: 1-bromo-2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 296.97
Molecular Formula: C7BrF7
InChi Key: DONVNRFILRLHJB-UHFFFAOYSA-N
SMILES: FC1=C(F)C(=C(F)C(F)=C1Br)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)