missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,2-diphenylbutyronitrile 95.0+%, TCI America™
Supplier: TCI America B1331250G
Specifications
| 4-Bromo-2,2-diphenylbutyronitrile | |
| 70°C | |
| C16H14BrN | |
| 250 g | |
| IGYSFJHVFHNOEI-UHFFFAOYSA-N | |
| 4-bromo-2,2-diphenylbutanenitrile | |
| 96575 | |
| ≥95.0% (GC) |
| 39186-58-8 | |
| White-Yellow | |
| MFCD00001845 | |
| 4-bromo-2,2-diphenylbutyronitrile, 2,2-diphenyl-4-bromobutyronitrile, 4-bromo-2,2-diphenyl-butyronitrile, 3-cyano-3,3-diphenylpropyl bromide, benzeneacetonitrile, .alpha.-2-bromoethyl-.alpha.-phenyl, acmc-1csza, butyronitrile,2-diphenyl, ksc495i2r, aronis25014, bromoethyl diphenyl acetonitrile | |
| C1=CC=C(C=C1)C(CCBr)(C#N)C2=CC=CC=C2 | |
| 300.199 | |
| 300.20 | |
| Crystalline Powder |
Chemical Identifiers
| 39186-58-8 | |
| 300.199 | |
| IGYSFJHVFHNOEI-UHFFFAOYSA-N | |
| 96575 | |
| C1=CC=C(C=C1)C(CCBr)(C#N)C2=CC=CC=C2 |
| C16H14BrN | |
| MFCD00001845 | |
| 4-bromo-2,2-diphenylbutyronitrile, 2,2-diphenyl-4-bromobutyronitrile, 4-bromo-2,2-diphenyl-butyronitrile, 3-cyano-3,3-diphenylpropyl bromide, benzeneacetonitrile, .alpha.-2-bromoethyl-.alpha.-phenyl, acmc-1csza, butyronitrile,2-diphenyl, ksc495i2r, aronis25014, bromoethyl diphenyl acetonitrile | |
| 4-bromo-2,2-diphenylbutanenitrile |
Safety and Handling
TSCA : No