missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,2-diphenylbutyronitrile 95.0+%, TCI America™

Catalog No. p-7118997
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B133125G 25 g
B1331250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B133125G Supplier TCI America Supplier No. B133125G
Only null left

Chemical Identifiers

CAS 39186-58-8
Molecular Formula C16H14BrN
Molecular Weight (g/mol) 300.199
MDL Number MFCD00001845
InChI Key IGYSFJHVFHNOEI-UHFFFAOYSA-N
Synonym 4-bromo-2,2-diphenylbutyronitrile, 2,2-diphenyl-4-bromobutyronitrile, 4-bromo-2,2-diphenyl-butyronitrile, 3-cyano-3,3-diphenylpropyl bromide, benzeneacetonitrile, .alpha.-2-bromoethyl-.alpha.-phenyl, acmc-1csza, butyronitrile,2-diphenyl, ksc495i2r, aronis25014, bromoethyl diphenyl acetonitrile
PubChem CID 96575
IUPAC Name 4-bromo-2,2-diphenylbutanenitrile
SMILES C1=CC=C(C=C1)C(CCBr)(C#N)C2=CC=CC=C2

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 25 g
Formula Weight 300.20
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,2-diphenylbutyronitrile