missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,2-diphenylbutyric Acid 98.0+%, TCI America™
Supplier: TCI America B359725G
Specifications
| 4-Bromo-2,2-diphenylbutyric Acid | |
| 127°C | |
| MFCD01321300 | |
| GFIYIIRFIODLLU-UHFFFAOYSA-N | |
| 4-bromo-2,2-diphenylbutanoic acid | |
| 2756959 | |
| ≥98.0% (T) |
| 37742-98-6 | |
| C16H15BrO2 | |
| 25 g | |
| C1=CC=C(C=C1)C(CCBr)(C2=CC=CC=C2)C(=O)O | |
| 319.198 | |
| 319.20 | |
| Crystalline Powder |
Chemical Identifiers
| 37742-98-6 | |
| 319.198 | |
| GFIYIIRFIODLLU-UHFFFAOYSA-N | |
| 4-bromo-2,2-diphenylbutanoic acid |
| C16H15BrO2 | |
| MFCD01321300 | |
| 2756959 | |
| C1=CC=C(C=C1)C(CCBr)(C2=CC=CC=C2)C(=O)O |
Safety and Handling
TSCA : No