missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,2-diphenylbutyric Acid 98.0+%, TCI America™

Catalog No. p-7123035
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35975G 5 g
B359725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35975G Supplier TCI America Supplier No. B35975G

Chemical Identifiers

CAS 37742-98-6
Molecular Formula C16H15BrO2
Molecular Weight (g/mol) 319.198
MDL Number MFCD01321300
InChI Key GFIYIIRFIODLLU-UHFFFAOYSA-N
PubChem CID 2756959
IUPAC Name 4-bromo-2,2-diphenylbutanoic acid
SMILES C1=CC=C(C=C1)C(CCBr)(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 127°C
Quantity 5 g
Formula Weight 319.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2,2-diphenylbutyric Acid