missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-indanone, 97%, Thermo Scientific™

Chemical Identifiers

CAS 15115-60-3
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.06
MDL Number MFCD01719772
InChI Key UVVYFYLSZIMKMC-UHFFFAOYSA-N
Synonym 4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone
PubChem CID 98713
IUPAC Name 4-bromo-2,3-dihydroinden-1-one
SMILES BrC1=CC=CC2=C1CCC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427700010
SDS View Documents
Thermo Scientific Chemicals
427700010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC427700050
SDS View Documents
Thermo Scientific Chemicals
427700050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

15115-60-3
211.06
UVVYFYLSZIMKMC-UHFFFAOYSA-N
98713
BrC1=CC=CC2=C1CCC2=O
C9H7BrO
MFCD01719772
4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone
4-bromo-2,3-dihydroinden-1-one

Specifications

15115-60-3
100.0
96% min. (GC)
C9H7BrO
1g
UVVYFYLSZIMKMC-UHFFFAOYSA-N
4-bromo-2,3-dihydroinden-1-one
98713
97%
96.0
Authentic
Glass bottle
MFCD01719772
4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone
BrC1=CC=CC2=C1CCC2=O
211.06
211.05
4-Bromo-1-indanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only