missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-indanone, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 427700010

 View more versions of this product

Catalog No. AC427700010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Bromo-1-indanone
96.0
Authentic
Glass bottle
MFCD01719772
4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone
BrC1=CC=CC2=C1CCC2=O
211.06
211.05
15115-60-3
100.0
96% min. (GC)
C9H7BrO
1g
UVVYFYLSZIMKMC-UHFFFAOYSA-N
4-bromo-2,3-dihydroinden-1-one
98713
97%

Chemical Identifiers

15115-60-3
211.06
UVVYFYLSZIMKMC-UHFFFAOYSA-N
98713
BrC1=CC=CC2=C1CCC2=O
C9H7BrO
MFCD01719772
4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone
4-bromo-2,3-dihydroinden-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only