accessibility menu, dialog, popup

UserName

BrO

CAS RN 15115-60-3

IUPAC Name: 4-bromo-2,3-dihydro-1H-inden-1-one
Synonyms: 4-bromo-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 211.06
Molecular Formula: C9H7BrO
InChi Key: UVVYFYLSZIMKMC-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1CCC2=O
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)