missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-indanone, 97%

CAS: 15115-60-3 | C9H7BrO | 211.06 g/mol

Supplier:  Thermo Scientific Chemicals H5693706

 View more versions of this product

Catalog No. AAH5693706


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-1-indanone
95°C to 99°C
MFCD01719772
4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone
BrC1=CC=CC2=C1CCC2=O
211.06
211.06
15115-60-3
C9H7BrO
5 g
UVVYFYLSZIMKMC-UHFFFAOYSA-N
4-bromo-2,3-dihydroinden-1-one
98713
97%

Chemical Identifiers

15115-60-3
211.06
UVVYFYLSZIMKMC-UHFFFAOYSA-N
98713
BrC1=CC=CC2=C1CCC2=O
C9H7BrO
MFCD01719772
4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone
4-bromo-2,3-dihydroinden-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

RTECSNumber : NK7536100

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only