missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-indanone, 97%

Catalog No. p-7038055
Change view
Click to view available options
Quantity:
5 g
10 g
Catalog No. Quantity
AAH5693706 5 g
AAH5693709 10 g
2 options

Catalog No. AAH5693706

Supplier: Thermo Scientific Chemicals H5693706

Only null left

CAS: 15115-60-3 | C9H7BrO | 211.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15115-60-3
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.06
MDL Number MFCD01719772
InChI Key UVVYFYLSZIMKMC-UHFFFAOYSA-N
Synonym 4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone
PubChem CID 98713
IUPAC Name 4-bromo-2,3-dihydroinden-1-one
SMILES BrC1=CC=CC2=C1CCC2=O

Specifications

Melting Point 95°C to 99°C
Quantity 5 g
Formula Weight 211.06
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-indanone
Hazard Category H315-H319
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
RTECSNumber NK7536100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only