missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-1-chloro-2-fluorobenzene 98.0+%, TCI America™
$52.19 - $205.05
Chemical Identifiers
| CAS | 60811-18-9 |
|---|---|
| Molecular Formula | C6H3BrClF |
| Molecular Weight (g/mol) | 209.44 |
| MDL Number | MFCD00042519 |
| InChI Key | AGYWDGVTLKNTBS-UHFFFAOYSA-N |
| Synonym | 1-bromo-4-chloro-3-fluorobenzene, 3-fluoro-4-chlorobromobenzene, 2-fluoro-4-bromochlorobenzene, 2-chloro-5-bromofluorobenzene, 4-chloro-3-fluorobromobenzene, 5-bromo-2-chlorofluorobenzene, 4-bromo-1-chloro-2-fluoro-benzene, 4-bromo-2-fluorochlorobenzene, benzene, 4-bromo-1-chloro-2-fluoro, 3-fluoro-4-chloro-bromobenzene |
| PubChem CID | 2724690 |
| IUPAC Name | 4-bromo-1-chloro-2-fluorobenzene |
| SMILES | FC1=C(Cl)C=CC(Br)=C1 |
Chemical Identifiers
| 60811-18-9 | |
| 209.44 | |
| AGYWDGVTLKNTBS-UHFFFAOYSA-N | |
| 2724690 | |
| FC1=C(Cl)C=CC(Br)=C1 |
| C6H3BrClF | |
| MFCD00042519 | |
| 1-bromo-4-chloro-3-fluorobenzene, 3-fluoro-4-chlorobromobenzene, 2-fluoro-4-bromochlorobenzene, 2-chloro-5-bromofluorobenzene, 4-chloro-3-fluorobromobenzene, 5-bromo-2-chlorofluorobenzene, 4-bromo-1-chloro-2-fluoro-benzene, 4-bromo-2-fluorochlorobenzene, benzene, 4-bromo-1-chloro-2-fluoro, 3-fluoro-4-chloro-bromobenzene | |
| 4-bromo-1-chloro-2-fluorobenzene |
Specifications
| 60811-18-9 | |
| 142°C | |
| MFCD00042519 | |
| 1-bromo-4-chloro-3-fluorobenzene, 3-fluoro-4-chlorobromobenzene, 2-fluoro-4-bromochlorobenzene, 2-chloro-5-bromofluorobenzene, 4-chloro-3-fluorobromobenzene, 5-bromo-2-chlorofluorobenzene, 4-bromo-1-chloro-2-fluoro-benzene, 4-bromo-2-fluorochlorobenzene, benzene, 4-bromo-1-chloro-2-fluoro, 3-fluoro-4-chloro-bromobenzene | |
| FC1=C(Cl)C=CC(Br)=C1 | |
| 209.44 | |
| 209.44 | |
| Liquid |
| Yellow | |
| C6H3BrClF | |
| 5 g | |
| AGYWDGVTLKNTBS-UHFFFAOYSA-N | |
| 4-bromo-1-chloro-2-fluorobenzene | |
| 2724690 | |
| ≥98.0% (GC) | |
| 4-Bromo-1-chloro-2-fluorobenzene |
Safety and Handling
TSCA : No