missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-chloro-2-fluorobenzene 98.0+%, TCI America™

Catalog No. B18615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18615G 5 g
B186125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18615G Supplier TCI America Supplier No. B18615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60811-18-9
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.44
MDL Number MFCD00042519
InChI Key AGYWDGVTLKNTBS-UHFFFAOYSA-N
Synonym 1-bromo-4-chloro-3-fluorobenzene, 3-fluoro-4-chlorobromobenzene, 2-fluoro-4-bromochlorobenzene, 2-chloro-5-bromofluorobenzene, 4-chloro-3-fluorobromobenzene, 5-bromo-2-chlorofluorobenzene, 4-bromo-1-chloro-2-fluoro-benzene, 4-bromo-2-fluorochlorobenzene, benzene, 4-bromo-1-chloro-2-fluoro, 3-fluoro-4-chloro-bromobenzene
PubChem CID 2724690
IUPAC Name 4-bromo-1-chloro-2-fluorobenzene
SMILES FC1=C(Cl)C=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 g
Formula Weight 209.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-chloro-2-fluorobenzene