missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-chloro-2-fluorobenzene, 98%

CAS: 60811-18-9 | C6H3BrClF | 209.44 g/mol

$197.71 - $543.71

Chemical Identifiers

CAS 60811-18-9
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.44
MDL Number MFCD00042519
InChI Key AGYWDGVTLKNTBS-UHFFFAOYSA-N
Synonym 1-bromo-4-chloro-3-fluorobenzene, 3-fluoro-4-chlorobromobenzene, 2-fluoro-4-bromochlorobenzene, 2-chloro-5-bromofluorobenzene, 4-chloro-3-fluorobromobenzene, 5-bromo-2-chlorofluorobenzene, 4-bromo-1-chloro-2-fluoro-benzene, 4-bromo-2-fluorochlorobenzene, benzene, 4-bromo-1-chloro-2-fluoro, 3-fluoro-4-chloro-bromobenzene
PubChem CID 2724690
IUPAC Name 4-bromo-1-chloro-2-fluorobenzene
SMILES FC1=C(Cl)C=CC(Br)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1574514
View Documents
Thermo Scientific Chemicals
A1574514
25 g
Each for $197.71
Only null left
 
AAA1574522
View Documents
Thermo Scientific Chemicals
A1574522
100 g
Each for $543.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

60811-18-9
209.44
AGYWDGVTLKNTBS-UHFFFAOYSA-N
2724690
FC1=C(Cl)C=CC(Br)=C1
C6H3BrClF
MFCD00042519
1-bromo-4-chloro-3-fluorobenzene, 3-fluoro-4-chlorobromobenzene, 2-fluoro-4-bromochlorobenzene, 2-chloro-5-bromofluorobenzene, 4-chloro-3-fluorobromobenzene, 5-bromo-2-chlorofluorobenzene, 4-bromo-1-chloro-2-fluoro-benzene, 4-bromo-2-fluorochlorobenzene, benzene, 4-bromo-1-chloro-2-fluoro, 3-fluoro-4-chloro-bromobenzene
4-bromo-1-chloro-2-fluorobenzene

Specifications

60811-18-9
62°C to 64°C (9 mmHg)
1.555
25 g
1-bromo-4-chloro-3-fluorobenzene, 3-fluoro-4-chlorobromobenzene, 2-fluoro-4-bromochlorobenzene, 2-chloro-5-bromofluorobenzene, 4-chloro-3-fluorobromobenzene, 5-bromo-2-chlorofluorobenzene, 4-bromo-1-chloro-2-fluoro-benzene, 4-bromo-2-fluorochlorobenzene, benzene, 4-bromo-1-chloro-2-fluoro, 3-fluoro-4-chloro-bromobenzene
FC1=C(Cl)C=CC(Br)=C1
209.44
209.45
4-Bromo-1-chloro-2-fluorobenzene
1.71
C6H3BrClF
MFCD00042519
2206649
AGYWDGVTLKNTBS-UHFFFAOYSA-N
4-bromo-1-chloro-2-fluorobenzene
2724690
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only