missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-1-butanol (contains varying amounts of Tetrahydrofuran) 80.0+%, TCI America™
Supplier: TCI America B188525G
Specifications
| 4-Bromo-1-butanol (contains varying amounts of Tetrahydrofuran) | |
| Yellow | |
| MFCD00015387 | |
| 1993 | |
| SIJLYRDVTMMSIP-UHFFFAOYSA-N | |
| 4-bromobutan-1-ol | |
| 118388 | |
| ≥80.0% (NMR) |
| 33036-62-3 | |
| C4H9BrO | |
| 25 g | |
| 4-bromo-1-butanol, 1-butanol, 4-bromo, 4-bromobutanol, 4-bromo-butan-1-ol, 1-bromo-4-hydroxybutane, tetramethylene bromohydrin, 4-bromo-n-butanol, 1-bromobutan-4-ol, 4-bromo butan-1-ol, acmc-1axcv | |
| C(CCBr)CO | |
| 153.019 | |
| 153.02 | |
| Liquid |
Chemical Identifiers
| 33036-62-3 | |
| 153.019 | |
| SIJLYRDVTMMSIP-UHFFFAOYSA-N | |
| 118388 | |
| C(CCBr)CO |
| C4H9BrO | |
| MFCD00015387 | |
| 4-bromo-1-butanol, 1-butanol, 4-bromo, 4-bromobutanol, 4-bromo-butan-1-ol, 1-bromo-4-hydroxybutane, tetramethylene bromohydrin, 4-bromo-n-butanol, 1-bromobutan-4-ol, 4-bromo butan-1-ol, acmc-1axcv | |
| 4-bromobutan-1-ol |
Safety and Handling
EINECSNumber : (2)-2002
TSCA : No
Recommended Storage : Freezer