accessibility menu, dialog, popup

UserName

HOBr

CAS RN 33036-62-3

IUPAC Name: 4-bromobutan-1-ol
Synonyms: 4-bromobutan-1-ol
Molecular Weight (g/mol): 153.02
Molecular Formula: C4H9BrO
InChi Key: SIJLYRDVTMMSIP-UHFFFAOYSA-N
SMILES: OCCCCBr
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (5)
  • (3)
  • (8)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)