missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-butanol (contains varying amounts of Tetrahydrofuran) 80.0+%, TCI America™

Catalog No. p-7119329
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B18855G 5 g
B188525G 25 g
2 options

Catalog No. B18855G

Supplier: TCI America B18855G

Only null left

Chemical Identifiers

CAS 33036-62-3
Molecular Formula C4H9BrO
Molecular Weight (g/mol) 153.019
MDL Number MFCD00015387
InChI Key SIJLYRDVTMMSIP-UHFFFAOYSA-N
Synonym 4-bromo-1-butanol, 1-butanol, 4-bromo, 4-bromobutanol, 4-bromo-butan-1-ol, 1-bromo-4-hydroxybutane, tetramethylene bromohydrin, 4-bromo-n-butanol, 1-bromobutan-4-ol, 4-bromo butan-1-ol, acmc-1axcv
PubChem CID 118388
IUPAC Name 4-bromobutan-1-ol
SMILES C(CCBr)CO

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 153.02
Percent Purity ≥80.0% (NMR)
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-butanol (contains varying amounts of Tetrahydrofuran)
EINECSNumber (2)-2002
TSCA No
Recommended Storage Freezer