missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,8-naphthalic Anhydride 95.0+%, TCI America™

Catalog No. B08585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08585G 5 g
B085825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08585G Supplier TCI America Supplier No. B08585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81-86-7
Molecular Formula C12H5BrO3
Molecular Weight (g/mol) 277.073
MDL Number MFCD00006927
InChI Key DTUOTSLAFJCQHN-UHFFFAOYSA-N
Synonym 4-Bromo-1,8-naphthalenedicarboxylic Anhydride
PubChem CID 66493
SMILES C1=CC2=C(C=CC3=C2C(=C1)C(=O)OC3=O)Br

Specifications

Melting Point 221°C
Color White-Yellow
Quantity 5 g
Formula Weight 277.07
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-1,8-naphthalic Anhydride