accessibility menu, dialog, popup

UserName

CAS RN 81-86-7

IUPAC Name: 10-bromo-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Synonyms: 10-bromo-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Molecular Weight (g/mol): 277.07
Molecular Formula: C12H5BrO3
InChi Key: DTUOTSLAFJCQHN-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=C3C(=O)OC(=O)C(C=C1)=C23
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results