missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,2-dichlorobenzene, 98+%

CAS: 18282-59-2 | C6H3BrCl2 | 225.89 g/mol

Supplier:  Thermo Scientific Chemicals A1031414

 View more versions of this product

Catalog No. AAA1031414

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-1,2-dichlorobenzene
22°C to 25°C
140°C to 142°C (30 mmHg)
C6H3BrCl2
25 g
1-bromo-3,4-dichlorobenzene, 3,4-dichlorobromobenzene, benzene, 4-bromo-1,2-dichloro, 3,4-dichlorophenyl bromide, 3,4-dichloro-1-bromobenzene, 4-bromo-1,2-dichloro-benzene, 5-bromdichlorbenzol, pubchem3611, acmc-1btol, 3,4-dichloro-bromobenzene
ClC1=CC=C(Br)C=C1Cl
225.89
225.91
18282-59-2
1.781
87°C (188°F)
MFCD00040849
1635735
CFPZDVAZISWERM-UHFFFAOYSA-N
4-bromo-1,2-dichlorobenzene
29013
≥98%

Chemical Identifiers

18282-59-2
225.89
CFPZDVAZISWERM-UHFFFAOYSA-N
29013
ClC1=CC=C(Br)C=C1Cl
C6H3BrCl2
MFCD00040849
1-bromo-3,4-dichlorobenzene, 3,4-dichlorobromobenzene, benzene, 4-bromo-1,2-dichloro, 3,4-dichlorophenyl bromide, 3,4-dichloro-1-bromobenzene, 4-bromo-1,2-dichloro-benzene, 5-bromdichlorbenzol, pubchem3611, acmc-1btol, 3,4-dichloro-bromobenzene
4-bromo-1,2-dichlorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 242-160-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only