accessibility menu, dialog, popup

UserName

CAS RN 18282-59-2

IUPAC Name: 4-bromo-1,2-dichlorobenzene
Synonyms: benzene, 4-bromo-1,2-dichloro-
Molecular Weight (g/mol): 225.89
Molecular Formula: C6H3BrCl2
InChi Key: CFPZDVAZISWERM-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Br)C=C1Cl
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)