missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacene 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 155°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 461.57 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Bromo-1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacene |
Safety and Handling
| TSCA | No |