accessibility menu, dialog, popup

UserName

H3CH3CH3CCH3CH3BrH3CH3CH3C

CAS RN 960079-28-1

IUPAC Name: 4-bromo-1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacene
Synonyms: 4-bromo-1,1,3,3,5,5,7,7-octaethyl-2,6-dihydro-s-indacene
Molecular Weight (g/mol): 461.57
Molecular Formula: C28H45Br
InChi Key: SNWPVKQFJCNSIU-UHFFFAOYSA-N
SMILES: CCC1(CC)CC(CC)(CC)C2=C(Br)C3=C(C=C12)C(CC)(CC)CC3(CC)CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results