missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacene 95.0+%, TCI America™

Catalog No. B43801G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43801G 1 g
B43805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43801G Supplier TCI America Supplier No. B43801G
Only null left

Chemical Identifiers

CAS 960079-28-1
Molecular Formula C28H45Br
Molecular Weight (g/mol) 461.572
InChI Key SNWPVKQFJCNSIU-UHFFFAOYSA-N
Synonym Eind-Br
PubChem CID 91972074
IUPAC Name 4-bromo-1,1,3,3,5,5,7,7-octaethyl-2,6-dihydro-s-indacene
SMILES CCC1(CC(C2=C(C3=C(C=C21)C(CC3(CC)CC)(CC)CC)Br)(CC)CC)CC

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 1 g
Formula Weight 461.57
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-1,1,3,3,5,5,7,7-octaethyl-1,2,3,5,6,7-hexahydro-s-indacene