missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. p-7039779
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59194MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH59194MD Supplier Thermo Scientific Chemicals Supplier No. H59194MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is the key intermediate in the synthesis of ,3-Dihydro-1,4-benzodiazepin-2(2H)-ones and 3-Amino-1,3-dihydro-1,4-benzodiazepin-2(2H)-ones. This synthesis is done with Pd-catalyzed cross-coupling of imidoyl chlorides with organoboronic acids.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 1256256-45-7
Molecular Formula C17H25BFNO4
Molecular Weight (g/mol) 337.198
MDL Number MFCD18383441
InChI Key FALNYBWTRBHWID-UHFFFAOYSA-N
Synonym tert-butyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl n-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino-2-fluorobenzeneboronic acid pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl phenyl carbamic acid tert-butyl ester
PubChem CID 59578631
IUPAC Name tert-butyl N-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)NC(=O)OC(C)(C)C)F

Specifications

Melting Point 150°C to 152°C
Quantity 250 mg
Formula Weight 337.2
Percent Purity 96%
Chemical Name or Material 4-(Boc-amino)-2-fluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures