missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylacetonitrile 95.0+%, TCI America™

Catalog No. B15455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15455G 5 g
B154525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15455G Supplier TCI America Supplier No. B15455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31603-77-7
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00016403
InChI Key HSZCNGTZJWZAMF-UHFFFAOYSA-N
Synonym 4-biphenylacetonitrile, 2-1,1'-biphenyl-4-yl acetonitrile, 2-4-phenylphenyl acetonitrile, 1,1'-biphenyl-4-acetonitrile, 4-cyanomethylbiphenyl, p-biphenylacetonitrile, p-biphenylylacetonitrile, 4-biphenyl-acetonitrile, 4-biphenylylacetonitrile, p-phenyl benzyl cyanide
PubChem CID 35856
IUPAC Name 2-{[1,1'-biphenyl]-4-yl}acetonitrile
SMILES N#CCC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 5 g
Formula Weight 193.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Biphenylacetonitrile