accessibility menu, dialog, popup

UserName

N

CAS RN 31603-77-7

IUPAC Name: 2-{[1,1'-biphenyl]-4-yl}acetonitrile
Synonyms: 2-{[1,1'-biphenyl]-4-yl}acetonitrile
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: HSZCNGTZJWZAMF-UHFFFAOYSA-N
SMILES: N#CCC1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)