missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzylthiomorpholine 1,1-Dioxide 98.0+%, TCI America™
$80.85 - $240.64
Chemical Identifiers
| CAS | 26475-66-1 |
|---|---|
| Molecular Formula | C11H15NO2S |
| Molecular Weight (g/mol) | 225.31 |
| MDL Number | MFCD00084894 |
| InChI Key | KFAMTQFKYUXQKV-UHFFFAOYSA-N |
| PubChem CID | 318454 |
| IUPAC Name | 4-benzyl-1λâ¶-thiomorpholine-1,1-dione |
| SMILES | O=S1(=O)CCN(CC2=CC=CC=C2)CC1 |
Chemical Identifiers
| 26475-66-1 | |
| 225.31 | |
| KFAMTQFKYUXQKV-UHFFFAOYSA-N | |
| 4-benzyl-1λâ¶-thiomorpholine-1,1-dione |
| C11H15NO2S | |
| MFCD00084894 | |
| 318454 | |
| O=S1(=O)CCN(CC2=CC=CC=C2)CC1 |
Specifications
| 26475-66-1 | |
| C11H15NO2S | |
| 5 g | |
| KFAMTQFKYUXQKV-UHFFFAOYSA-N | |
| 4-benzyl-1λâ¶-thiomorpholine-1,1-dione | |
| 318454 | |
| ≥98.0% (GC) | |
| 4-Benzylthiomorpholine 1,1-Dioxide |
| White | |
| MFCD00084894 | |
| 4-benzylthiomorpholine 1,1-dioxide, 1,1-dioxide-4-phenylmethyl thiomorpholine, 4-benzyl-1??-thiomorpholine-1,1-dione, 4-benzylthiomorpholine1,1-dioxide, acmc-209grq, maybridge1_003205, 4-benzylthiomorpholine-1,1-dioxide, 4-benzylthiomorpholine-1,1,-dioxide, 4-benzyl-1,4-thiazaperhydroine-1,1-dione | |
| O=S1(=O)CCN(CC2=CC=CC=C2)CC1 | |
| 225.31 | |
| 225.31 | |
| Crystalline Powder |
Safety and Handling
TSCA : No