accessibility menu, dialog, popup

UserName

OSON

CAS RN 26475-66-1

IUPAC Name: 4-benzyl-1λ⁶-thiomorpholine-1,1-dione
Synonyms: 4-benzyl-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 225.31
Molecular Formula: C11H15NO2S
InChi Key: KFAMTQFKYUXQKV-UHFFFAOYSA-N
SMILES: O=S1(=O)CCN(CC2=CC=CC=C2)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results