missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzylthiomorpholine 1,1-Dioxide 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Synonym | 4-benzylthiomorpholine 1,1-dioxide, 1,1-dioxide-4-phenylmethyl thiomorpholine, 4-benzyl-1??-thiomorpholine-1,1-dione, 4-benzylthiomorpholine1,1-dioxide, acmc-209grq, maybridge1_003205, 4-benzylthiomorpholine-1,1-dioxide, 4-benzylthiomorpholine-1,1,-dioxide, 4-benzyl-1,4-thiazaperhydroine-1,1-dione |
| Formula Weight | 225.31 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Benzylthiomorpholine 1,1-Dioxide |
Safety and Handling
| TSCA | No |