Learn More
4-Benzyloxyphenol, 98+%
CAS: 103-16-2 | C13H12O2 | 200.237 g/mol
Supplier: Thermo Scientific Chemicals A148430B
Description
4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.
Solubility
Slightly soluble in water. Soluble in alcohol, acetone, benzene, and diethyl ether.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Specifications
| 4-Benzyloxyphenol | |
| 118°C to 122°C | |
| C13H12O2 | |
| 1000 g | |
| 14,6248 | |
| Slightly soluble in water. Soluble in alcohol,acetone,benzene,and diethyl ether. | |
| C1=CC=C(C=C1)COC2=CC=C(C=C2)O | |
| 200.237 | |
| CHEBI:34380 | |
| ≥98% |
| 103-16-2 | |
| 1.26 | |
| MFCD00002333 | |
| 1958305 | |
| 4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon | |
| VYQNWZOUAUKGHI-UHFFFAOYSA-N | |
| 4-phenylmethoxyphenol | |
| 7638 | |
| 200.24 |
Chemical Identifiers
| 103-16-2 | |
| MFCD00002333 | |
| 4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon | |
| CHEBI:34380 |
| C13H12O2 | |
| VYQNWZOUAUKGHI-UHFFFAOYSA-N | |
| 7638 | |
| 4-phenylmethoxyphenol |
Safety and Handling
GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c
H317-H319
EINECSNumber : 203-083-3
RTECSNumber : SJ7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only