accessibility menu, dialog, popup

UserName

CAS RN 103-16-2

IUPAC Name: 4-(benzyloxy)phenol
Synonyms: monobenzone MBEH monobenzyl ether of hydroquinone agerite monobenzon monobenzonum superlite (antioxidant) hydroquinone monobenzyl ether (substance) dermochinona P-(benzyloxy) phenol
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: VYQNWZOUAUKGHI-UHFFFAOYSA-N
SMILES: OC1=CC=C(OCC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)