missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzyl Alcohol 95.0+%, TCI America™

Catalog No. p-7119147
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16025G 5 g
B160225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16025G Supplier TCI America Supplier No. B16025G

Chemical Identifiers

CAS 836-43-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00004654
InChI Key OEBIVOHKFYSBPE-UHFFFAOYSA-N
Synonym 4-benzyloxybenzyl alcohol, 4-benzyloxy phenyl methanol, p-benzyloxybenzyl alcohol, 4-benzyloxybenzylalcohol, 4-benzyloxy benzyl alcohol, benzenemethanol, 4-phenylmethoxy, p-alkoxybenzyl alcohol resin, 4-phenylmethoxyphenyl methanol, argopore-wang
PubChem CID 70043
ChEBI CHEBI:29486
IUPAC Name [4-(benzyloxy)phenyl]methanol
SMILES OCC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 5 g
Formula Weight 214.26
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxybenzyl Alcohol