missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzyl alcohol, 98+%

Catalog No. p-7022260
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1494909 10 g
AAA1494918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1494909 Supplier Thermo Scientific Chemicals Supplier No. A1494909
Only null left

CAS: 836-43-1 | C14H14O2 | 214.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 836-43-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00004654
InChI Key OEBIVOHKFYSBPE-UHFFFAOYSA-N
Synonym 4-benzyloxybenzyl alcohol, 4-benzyloxy phenyl methanol, p-benzyloxybenzyl alcohol, 4-benzyloxybenzylalcohol, 4-benzyloxy benzyl alcohol, benzenemethanol, 4-phenylmethoxy, p-alkoxybenzyl alcohol resin, 4-phenylmethoxyphenyl methanol, argopore-wang
PubChem CID 70043
ChEBI CHEBI:29486
IUPAC Name (4-phenylmethoxyphenyl)methanol
SMILES OCC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 84°C to 88°C
Quantity 10 g
Beilstein 1877819
Formula Weight 214.26
Percent Purity ≥98%
Chemical Name or Material 4-Benzyloxybenzyl alcohol
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 212-649-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only