missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybenzophenone 98.0+%, TCI America™

Catalog No. B45045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45045G 5 g
B450425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45045G Supplier TCI America Supplier No. B45045G
Only null left

Chemical Identifiers

CAS 54589-41-2
Molecular Formula C20H16O2
Molecular Weight (g/mol) 288.346
MDL Number MFCD00017508
InChI Key NMPNTBQOLRXPGK-UHFFFAOYSA-N
Synonym 4-benzyloxybenzophenone, 4-benzyloxy benzophenone, 4-benzyloxy phenyl phenyl methanone, phenyl-4-phenylmethoxyphenyl methanone, phenyl 4-phenylmethoxy phenyl ketone, acmc-20amoo, 4-benzyl-oxybenzophenone, 4-benzyloxyphenyl-phenyl-methanone
PubChem CID 458193
IUPAC Name phenyl-(4-phenylmethoxyphenyl)methanone
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)C(=O)C3=CC=CC=C3

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 5 g
Formula Weight 288.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxybenzophenone