CAS RN 54589-41-2
IUPAC Name:
[4-(benzyloxy)phenyl](phenyl)methanone
Synonyms:
[4-(benzyloxy)phenyl](phenyl)methanone
Molecular Weight (g/mol):
288.35
Molecular Formula:
C20H16O2
InChi Key:
NMPNTBQOLRXPGK-UHFFFAOYSA-N
SMILES:
O=C(C1=CC=CC=C1)C1=CC=C(OCC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
4-Benzyloxybenzophenone 98.0+%, TCI America™
CAS: 54589-41-2 Molecular Formula: C20H16O2 Molecular Weight (g/mol): 288.346 MDL Number: MFCD00017508 InChI Key: NMPNTBQOLRXPGK-UHFFFAOYSA-N Synonym: 4-benzyloxybenzophenone,4-benzyloxy benzophenone,4-benzyloxy phenyl phenyl methanone,phenyl-4-phenylmethoxyphenyl methanone,phenyl 4-phenylmethoxy phenyl ketone,acmc-20amoo,4-benzyl-oxybenzophenone,4-benzyloxyphenyl-phenyl-methanone PubChem CID: 458193 IUPAC Name: phenyl-(4-phenylmethoxyphenyl)methanone SMILES: C1=CC=C(C=C1)COC2=CC=C(C=C2)C(=O)C3=CC=CC=C3