missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxy)phenol 99.0+%, TCI America™

Catalog No. B110425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B110425G 25 g
B1104100G 100 g
B1104500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B110425G Supplier TCI America Supplier No. B110425G

Chemical Identifiers

CAS 103-16-2
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00002333
InChI Key VYQNWZOUAUKGHI-UHFFFAOYSA-N
Synonym 4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon
PubChem CID 7638
ChEBI CHEBI:34380
IUPAC Name 4-phenylmethoxyphenol
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)O

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 25 g
Formula Weight 200.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Benzyloxy)phenol
EINECSNumber (3)-0657
RTECSNumber SJ7700000
TSCA Yes