missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzyloxy-3-methoxyphenylacetic Acid 98.0+%, TCI America™
$298.25 - $885.43
Chemical Identifiers
| CAS | 29973-91-9 |
|---|---|
| Molecular Formula | C16H16O4 |
| Molecular Weight (g/mol) | 272.3 |
| MDL Number | MFCD00185562 |
| InChI Key | AYCIHUSEBJLTBF-UHFFFAOYSA-N |
| PubChem CID | 297964 |
| IUPAC Name | 2-(3-methoxy-4-phenylmethoxyphenyl)acetic acid |
| SMILES | COC1=C(C=CC(=C1)CC(=O)O)OCC2=CC=CC=C2 |
Chemical Identifiers
| 29973-91-9 | |
| 272.3 | |
| AYCIHUSEBJLTBF-UHFFFAOYSA-N | |
| 2-(3-methoxy-4-phenylmethoxyphenyl)acetic acid |
| C16H16O4 | |
| MFCD00185562 | |
| 297964 | |
| COC1=C(C=CC(=C1)CC(=O)O)OCC2=CC=CC=C2 |
Specifications
| 29973-91-9 | |
| White-Yellow | |
| MFCD00185562 | |
| AYCIHUSEBJLTBF-UHFFFAOYSA-N | |
| 2-(3-methoxy-4-phenylmethoxyphenyl)acetic acid | |
| 297964 | |
| ≥98.0% (GC,T) | |
| 4-Benzyloxy-3-methoxyphenylacetic Acid |
| 118°C | |
| C16H16O4 | |
| 1 g | |
| COC1=C(C=CC(=C1)CC(=O)O)OCC2=CC=CC=C2 | |
| 272.3 | |
| 272.30 | |
| Crystalline Powder |
Safety and Handling
TSCA : No