accessibility menu, dialog, popup

UserName

H3COOHOO

CAS RN 29973-91-9

IUPAC Name: 2-[4-(benzyloxy)-3-methoxyphenyl]acetic acid
Synonyms: [4-(benzyloxy)-3-methoxyphenyl]acetic acid
Molecular Weight (g/mol): 272.3
Molecular Formula: C16H16O4
InChi Key: AYCIHUSEBJLTBF-UHFFFAOYSA-N
SMILES: COC1=CC(CC(O)=O)=CC=C1OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results