missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3,5-dimethylbenzaldehyde, 95%

Catalog No. p-7026203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2206703 1 g
AAB2206706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2206703 Supplier Thermo Scientific Chemicals Supplier No. B2206703

CAS: 144896-51-5 | C16H16O2 | 240.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144896-51-5
Molecular Formula C16H16O2
Molecular Weight (g/mol) 240.30
MDL Number MFCD00800683
InChI Key GSYUTKRSEZMBNC-UHFFFAOYSA-N
Synonym 4-benzyloxy-3,5-dimethylbenzaldehyde, 3,5-dimethyl-4-phenylmethoxy benzaldehyde, zlchem 1072, acmc-1bxy6, 3,5-dimethyl-4-benzyloxybenzaldehyde, 3,5-dimethyl-4-benzyloxy benzaldehyde, 4-benzyloxy-3,5-dimethyl-benzaldehyde
PubChem CID 563557
IUPAC Name 3,5-dimethyl-4-phenylmethoxybenzaldehyde
SMILES CC1=CC(C=O)=CC(C)=C1OCC1=CC=CC=C1

Specifications

Density 1.1
Quantity 1 g
Refractive Index 1.586
Boiling Point 162°C to 163°C (1 mmHg)
Formula Weight 240.3
Percent Purity 95%
Chemical Name or Material 4-Benzyloxy-3,5-dimethylbenzaldehyde
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only