accessibility menu, dialog, popup

UserName

CH3OH3CO

CAS RN 144896-51-5

IUPAC Name: 4-(benzyloxy)-3,5-dimethylbenzaldehyde
Synonyms: 4-(benzyloxy)-3,5-dimethylbenzaldehyde
Molecular Weight (g/mol): 240.3
Molecular Formula: C16H16O2
InChi Key: GSYUTKRSEZMBNC-UHFFFAOYSA-N
SMILES: CC1=CC(C=O)=CC(C)=C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)