missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-1-bromo-2-fluorobenzene, 98%

Catalog No. AAH3005306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3005306 5 g
AAH3005314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3005306 Supplier Thermo Scientific Chemicals Supplier No. H3005306

CAS: 185346-79-6 | C13H10BrFO | 281.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 185346-79-6
Molecular Formula C13H10BrFO
Molecular Weight (g/mol) 281.124
MDL Number MFCD09801041
InChI Key BYTJTXKQBSNGCL-UHFFFAOYSA-N
Synonym 4-benzyloxy-1-bromo-2-fluorobenzene, 4-benzyloxy-2-fluorobromobenzene, benzene,1-bromo-2-fluoro-4-phenylmethoxy, acmc-1c1vq, benzyloxy-4-bromo-3-fluorobenzene, 1-benzyloxy-4-bromo-3-fluorobenzene, benzyl 4-bromo-3-fluorophenyl ether, 1-bromo-2-fluoro-4-benzyloxy benzene
PubChem CID 22273933
IUPAC Name 1-bromo-2-fluoro-4-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC(=C(C=C2)Br)F

Specifications

Melting Point 33°C to 35°C
Quantity 5 g
Formula Weight 281.13
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-1-bromo-2-fluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only