missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoyl-4'-bromobiphenyl 98.0+%, TCI America™

Catalog No. B272025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B272025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B272025G Supplier TCI America Supplier No. B272025G
Only null left

Chemical Identifiers

CAS 63242-14-8
Molecular Formula C19H13BrO
Molecular Weight (g/mol) 337.216
InChI Key ITMLYLPKSFZCJK-UHFFFAOYSA-N
Synonym 4-benzoyl-4'-bromobiphenyl, 4-4-bromophenyl benzophenone, 4'-bromobiphenyl-4-yl phenyl methanone, 4'-bromo-1,1'-biphenyl-4-yl phenyl methanone, 4-4-bromophenyl phenyl-phenyl-methanone, 4-4-bromophenyl phenyl phenyl ketone, 4-4-bromophenyl phenyl-phenylmethanone, 4-4-bromophenyl phenyl phenyl methanone, pubchem9089
PubChem CID 478901
IUPAC Name [4-(4-bromophenyl)phenyl]-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C3=CC=C(C=C3)Br

Specifications

Melting Point 157°C
Quantity 25 g
Formula Weight 337.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzoyl-4′-bromobiphenyl