accessibility menu, dialog, popup

UserName

BrO

CAS RN 63242-14-8

IUPAC Name: {4'-bromo-[1,1'-biphenyl]-4-yl}(phenyl)methanone
Synonyms: {4'-bromo-[1,1'-biphenyl]-4-yl}(phenyl)methanone
Molecular Weight (g/mol): 337.22
Molecular Formula: C19H13BrO
InChi Key: ITMLYLPKSFZCJK-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results